Assay Details
| Gene Symbol: EAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192153.3 | NP_001179082.2 | 5 - 6 | 1017 | 56 | |
| GenBank mRNA | BC149556.1 | - | 5 - 6 | 901 | 56 | |
More Information
Set Membership: |
|

EAF1
Genomic Map