Assay Details
| Gene Symbol: C1RL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001354760.1 | NP_001341689.1 | 3 - 4 | 551 | 59 | |
| GenBank mRNA | EV615074.1 | - | 2 - 3 | 270 | 59 | |
More Information
Set Membership: |
|

C1RL
Genomic Map