Assay Details
Gene Symbol: TNFAIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192170.1 | NP_001179099.1 | 2 - 3 | 481 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
