Assay Details
Gene Symbol: PLCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192255.2 | NP_001179184.2 | 1 - 2 | 305 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
