Assay Details
| Gene Symbol: PES1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001354818.1 | NP_001341747.1 | 6 - 7 | 716 | 58 | |
| GenBank mRNA | BC118072.1 | - | 5 - 6 | 2057 | 58 | |
| DT853420.1 | - | 6 - 7 | 651 | 58 | ||
| EH377693.1 | - | 6 - 7 | 652 | 58 | ||
More Information
Set Membership: |
|

PES1
Genomic Map