Assay Details
Gene Symbol: RCC1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192346.1 | NP_001179275.1 | 10 - 11 | 1353 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RCC1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192346.1 | NP_001179275.1 | 10 - 11 | 1353 | 76 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |