Assay Details
| Gene Symbol: ZNF384 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192394.1 | NP_001179323.1 | 7 - 8 | 1221 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF384| Gene Symbol: ZNF384 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192394.1 | NP_001179323.1 | 7 - 8 | 1221 | 64 | |
Set Membership: |
|