Assay Details
Gene Symbol: GNA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192401.3 | NP_001179330.1 | 2 - 3 | 599 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
