Assay Details
| Gene Symbol: FABP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192410.1 | NP_001179339.1 | 1 - 2 | 101 | 92 | |
| GenBank mRNA | CF768059.1 | - | 1 - 2 | 101 | 92 | |
More Information
Set Membership: |
|

FABP9
Genomic Map