Assay Details
| Gene Symbol: TM6SF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192430.1 | NP_001179359.1 | 1 - 2 | 199 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TM6SF2| Gene Symbol: TM6SF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192430.1 | NP_001179359.1 | 1 - 2 | 199 | 69 | |
Set Membership: |
|