Assay Details
| Gene Symbol: ACVR1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192879.2 | NP_001179808.2 | 4 - 5 | 1013 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ACVR1C| Gene Symbol: ACVR1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192879.2 | NP_001179808.2 | 4 - 5 | 1013 | 65 | |
Set Membership: |
|