Assay Details
| Gene Symbol: JPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192887.3 | NP_001179816.1 | 2 - 3 | 1142 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

JPH1| Gene Symbol: JPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192887.3 | NP_001179816.1 | 2 - 3 | 1142 | 72 | |
Set Membership: |
|