Assay Details
Gene Symbol: ZMPSTE24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192928.2 | NP_001179857.1 | 5 - 6 | 624 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZMPSTE24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192928.2 | NP_001179857.1 | 5 - 6 | 624 | 63 |
Set Membership: |
![]() ![]() ![]() ![]() |