Assay Details
Gene Symbol: RGS18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192971.1 | NP_001179900.1 | 2 - 3 | 399 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
