Assay Details
| Gene Symbol: DYM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192999.1 | NP_001179928.1 | 4 - 5 | 606 | 68 | |
| GenBank mRNA | BC105208.1 | - | 3 - 4 | 398 | 68 | |
More Information
Set Membership: |
|

DYM
Genomic Map