Assay Details
Gene Symbol: MEPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193004.3 | NP_001179933.3 | 4 - 5 | 232 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MEPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193004.3 | NP_001179933.3 | 4 - 5 | 232 | 98 |
Set Membership: |
![]() ![]() ![]() ![]() |