Assay Details
Gene Symbol: ZNF609 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193023.2 | NP_001179952.1 | 7 - 8 | 4148 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF609 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193023.2 | NP_001179952.1 | 7 - 8 | 4148 | 77 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |