Assay Details
| Gene Symbol: NUBPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193042.1 | NP_001179971.1 | 3 - 4 | 295 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NUBPL| Gene Symbol: NUBPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193042.1 | NP_001179971.1 | 3 - 4 | 295 | 73 | |
Set Membership: |
|