Assay Details
| Gene Symbol: HEXIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193043.1 | NP_001179972.1 | 2 - 3 | 260 | 107 | |
| GenBank mRNA | AB196139.1 | - | 2 - 3 | 272 | 107 | |
More Information
Set Membership: |
|

HEXIM2
Genomic Map