Assay Details
Gene Symbol: FEZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193050.2 | NP_001179979.1 | 4 - 5 | 643 | 74 | |
NM_001205335.1 | NP_001192264.1 | 4 - 5 | 643 | 74 | ||
GenBank mRNA | EE334609.1 | - | 4 - 5 | 620 | 74 | |
EH146291.1 | - | 3 - 4 | 534 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
