Assay Details
| Gene Symbol: VSIG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193134.2 | NP_001180063.2 | 2 - 3 | 226 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

VSIG2| Gene Symbol: VSIG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193134.2 | NP_001180063.2 | 2 - 3 | 226 | 63 | |
Set Membership: |
|