Assay Details
Gene Symbol: TC2N | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193204.1 | NP_001180133.1 | 4 - 5 | 651 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TC2N | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193204.1 | NP_001180133.1 | 4 - 5 | 651 | 71 |
Set Membership: |
![]() ![]() ![]() ![]() |