Assay Details
| Gene Symbol: EIF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193223.1 | NP_001180152.1 | 6 - 7 | 730 | 119 | |
| GenBank mRNA | BC154391.1 | - | 1 - 2 | 49 | 119 | |
More Information
Set Membership: |
|

EIF5
Genomic Map