Assay Details
Gene Symbol: ZFHX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193227.2 | NP_001180156.2 | 9 - 10 | 4091 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
