Assay Details
| Gene Symbol: CCR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194960.1 | NP_001181889.1 | 1 | 1021 | 86 | |
| GenBank mRNA | GU936961.1 | - | 1 | 1021 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCR3| Gene Symbol: CCR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194960.1 | NP_001181889.1 | 1 | 1021 | 86 | |
| GenBank mRNA | GU936961.1 | - | 1 | 1021 | 86 | |
Set Membership: |
|