Assay Details
Gene Symbol: ZMYM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195018.1 | NP_001181947.1 | 7 - 8 | 1535 | 105 | |
GenBank mRNA | BC116071.1 | - | 1 - 2 | 191 | 105 | |
BC149331.1 | - | 7 - 8 | 1536 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
