Assay Details
Gene Symbol: BOLA-DYB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012679.1 | NP_001012697.1 | 1 - 2 | 148 | 94 | |
GenBank mRNA | AJ580584.1 | - | 1 - 2 | 148 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
