Assay Details
Gene Symbol: BOLA-DOB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013600.1 | NP_001013618.1 | 2 - 3 | 503 | 85 | |
GenBank mRNA | AB117945.1 | - | 2 - 3 | 364 | 85 | |
AB117946.1 | - | 2 - 3 | 503 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
