Assay Details
| Gene Symbol: ARPC1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014844.3 | NP_001014844.1 | 9 - 10 | 1132 | 55 | |
| GenBank mRNA | BC102942.1 | - | 9 - 10 | 1133 | 55 | |
| BT021526.1 | - | 8 - 9 | 1085 | 55 | ||
| BT021895.1 | - | 8 - 9 | 1079 | 55 | ||
| BT025344.1 | - | 8 - 9 | 1073 | 55 | ||
| BT026160.1 | - | 8 - 9 | 1074 | 55 | ||
| BT026274.1 | - | 8 - 9 | 1073 | 55 | ||
More Information
Set Membership: |
|

ARPC1B
Genomic Map