Assay Details
Gene Symbol: SSNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015539.1 | NP_001015539.1 | 2 - 3 | 275 | 85 | |
GenBank mRNA | BC111237.1 | - | 2 - 3 | 255 | 85 | |
BT020997.1 | - | 2 - 3 | 275 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
