Assay Details
| Gene Symbol: DHX58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015545.1 | NP_001015545.1 | 10 - 11 | 1793 | 66 | |
| GenBank mRNA | BT020952.1 | - | 10 - 11 | 1793 | 66 | |
More Information
Set Membership: |
|

DHX58
Genomic Map