Assay Details
| Gene Symbol: CRIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025349.2 | NP_001020520.1 | 197 | 96 | ||
| GenBank mRNA | AY911323.1 | - | 2 - 3 | 199 | 96 | |
| BC102754.1 | - | 197 | 96 | |||
More Information
Set Membership: |
|

CRIP1
Genomic Map