Assay Details
| Gene Symbol: CRIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025349.2 | NP_001020520.1 | 248 | 104 | ||
| GenBank mRNA | AY911323.1 | - | 3 - 4 | 250 | 104 | |
| BC102754.1 | - | 248 | 104 | |||
More Information
Set Membership: |
|

CRIP1
Genomic Map