Assay Details
Gene Symbol: NDUFB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033620.2 | NP_001028792.1 | 2 - 3 | 382 | 83 | |
GenBank mRNA | AJ510149.1 | - | 2 - 3 | 277 | 83 | |
BC102141.1 | - | 2 - 3 | 383 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
