Assay Details
| Gene Symbol: MRO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034552.1 | NP_001029724.1 | 2 - 3 | 140 | 105 | |
| GenBank mRNA | BC146146.1 | - | 2 - 3 | 246 | 105 | |
| BT021655.1 | - | 2 - 3 | 140 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC146146.1, NM_001034552.1 transcripts.
Set Membership: |
|

MRO
Genomic Map