Assay Details
| Gene Symbol: LOC781423 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037490.1 | NP_001032567.1 | 2 - 3 | 273 | 101 | |
| NM_001105012.2 | NP_001098482.1 | 3 - 4 | 1575 | 101 | ||
| GenBank mRNA | BC108236.1 | - | 2 - 3 | 273 | 101 | |
| BC149901.1 | - | 3 - 4 | 1576 | 101 | ||
More Information
Set Membership: |
|

LOC781423
Genomic Map