Assay Details
| Gene Symbol: PRODH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075185.1 | NP_001068653.1 | 4 - 5 | 673 | 71 | |
| GenBank mRNA | BC118353.1 | - | 4 - 5 | 673 | 71 | |
More Information
Set Membership: |
|

PRODH
Genomic Map