Assay Details
| Gene Symbol: PRKAR1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075201.1 | NP_001068669.1 | 8 - 9 | 824 | 63 | |
| GenBank mRNA | BC118392.1 | - | 8 - 9 | 824 | 63 | |
More Information
Set Membership: |
|

PRKAR1B
Genomic Map