Assay Details
Gene Symbol: GRM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076055.3 | NP_001069523.3 | 2 - 3 | 960 | 61 | |
GenBank mRNA | DV880013.1 | - | 1 - 2 | 212 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
