Assay Details
| Gene Symbol: TNNC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076373.1 | NP_001069841.1 | 2 - 3 | 103 | 69 | |
| GenBank mRNA | BC118491.1 | - | 2 - 3 | 103 | 69 | |
More Information
Set Membership: |
|

TNNC2
Genomic Map