Assay Details
| Gene Symbol: YAE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199002.1 | NP_001185931.1 | 1 - 2 | 193 | 78 | |
| GenBank mRNA | BC118387.1 | - | 1 - 2 | 194 | 78 | |
| EH154864.1 | - | 1 - 2 | 165 | 78 | ||
| EV742064.1 | - | 2 - 3 | 751 | 78 | ||
More Information
Set Membership: |
|

YAE1
Genomic Map