Assay Details
| Gene Symbol: RGS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199063.2 | NP_001185992.1 | 4 - 5 | 501 | 90 | |
| GenBank mRNA | BC149027.1 | - | 2 - 3 | 2205 | 90 | |
More Information
Set Membership: |
|

RGS1
Genomic Map