Assay Details
Gene Symbol: ZNF706 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199073.1 | NP_001186002.1 | 1 - 2 | 27 | 108 | |
GenBank mRNA | BC108246.1 | - | 1 - 2 | 29 | 108 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC108246.1, NM_001199073.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
