Assay Details
Gene Symbol: ZNF706 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199073.1 | NP_001186002.1 | 2 - 3 | 170 | 80 | |
NM_001199074.1 | NP_001186003.1 | 2 - 3 | 409 | 80 | ||
GenBank mRNA | BC108246.1 | - | 2 - 3 | 172 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
