Assay Details
Gene Symbol: ZNF706 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199073.1 | NP_001186002.1 | 3 - 4 | 278 | 112 | |
NM_001199074.1 | NP_001186003.1 | 3 - 4 | 517 | 112 | ||
GenBank mRNA | BC108246.1 | - | 3 - 4 | 280 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
