Assay Details
| Gene Symbol: C21H14orf180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205388.1 | NP_001192317.1 | 1 - 2 | 298 | 124 | |
| GenBank mRNA | BC114110.1 | - | 1 - 2 | 299 | 124 | |
| DV844194.1 | - | 1 - 2 | 299 | 124 | ||
| DV844417.1 | - | 1 - 2 | 298 | 124 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC114110.1, NM_001205388.1 transcripts.
Set Membership: |
|

C21H14orf180
Genomic Map