Assay Details
Gene Symbol: PRP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038132.1 | NP_001033221.1 | 2 - 3 | 275 | 105 | |
GenBank mRNA | AB204881.1 | - | 2 - 3 | 275 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
