Assay Details
| Gene Symbol: MT1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040492.2 | NP_001035582.1 | 1 - 2 | 104 | 118 | |
| GenBank mRNA | BC142364.1 | - | 1 - 2 | 102 | 118 | |
| DT859856.1 | - | 104 | 117 | |||
| DT862167.1 | - | 1 - 2 | 102 | 118 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC142364.1, NM_001040492.2 transcripts.
Set Membership: |
|

MT1A
Genomic Map