Assay Details
| Gene Symbol: NAA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046511.2 | NP_001039976.1 | 1 - 2 | 123 | 100 | |
| GenBank mRNA | BC112807.1 | - | 1 - 2 | 124 | 100 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC112807.1, NM_001046511.2 transcripts.
Set Membership: |
|

NAA10
Genomic Map