Assay Details
Gene Symbol: CRTAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206045.1 | NP_001192974.1 | 6 - 7 | 1444 | 124 | |
GenBank mRNA | BC109994.1 | - | 6 - 7 | 1196 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
